Order Entry
Puerto Rico
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
Catalog # 89139-118
Supplier:  Biotium
CAS Number:  
undefined
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
Catalog # 89139-118
Supplier:  Biotium
Supplier Number:  40043
CAS Number:  

Specifications

  • Size:
    1 ml
  • Cat. No.:
    89139-118
  • Supplier no.:
    40043

Specifications

About this item

 :
 :350.25 g/mol
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681