Order Entry
Puerto Rico
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) fluorescent dye, used in nuclear staining
Irritant
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) fluorescent dye, used in nuclear staining
Catalog # 89139-050
Supplier:  Biotium
CAS Number:  
Irritant
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) fluorescent dye, used in nuclear staining
Catalog # 89139-050
Supplier:  Biotium
Supplier Number:  40009
CAS Number:  

Specifications

  • Size:
    10 mg
  • Cat. No.:
    89139-050
  • Supplier no.:
    40009

Specifications

About this item

 :
 :457.48 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·₂C₃H₆O₃