Water with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
:64-18-6
:7732-18-5
:MFCD00011332
:0.998 g/cm³ (20 °C)
:Ambient
- Assay of formic acid:0.093 - 0.107 %
- pH (20 °C):2.6 - 2.8
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 210 nm:Min. 5 %
- Transmittance at 230 nm:Min. 45 %
- Transmittance at 254 nm:Min. 98 %
- HPLC Gradient at 210 nm:Max. 50 mAU
- HPLC Gradient at 254 nm:Max. 10 mAU
- LC-MS application:complying