Order Entry
Puerto Rico
ContactUsLinkComponent
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 89139-212
Supplier:  Biotium
CAS Number:  
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 89139-212
Supplier:  Biotium
Supplier Number:  50035
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89139-212
  • Supplier no.:
    50035

Specifications

About this item

 :
 :587 g/mol
Storage Temperature:Freezer
CAS Number:132319-57-4
Formula:C₁₈H₁₀K₄N₂O₁₁