Rhod-2 trisodium salt ≥90% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 89139-192
Supplier: Biotium
CAS Number:
Specifications
- Size:1 mg
- Cat. No.:89139-192
- Supplier no.:50022
Specifications
About this item
:
:945 g/mol
Storage Temperature:Refrigerator
Formula:C₄₀H₃₉N₄Na₃O₁₁