Order Entry
Puerto Rico
ContactUsLinkComponent
Pluronic® F-127 10% in water
Pluronic® F-127 10% in water
Catalog # 89139-290
Supplier:  Biotium
CAS Number:  
Pluronic® F-127 10% in water
Catalog # 89139-290
Supplier:  Biotium
Supplier Number:  59005
CAS Number:  

Specifications

  • Size:
    30 ml
  • Cat. No.:
    89139-290
  • Supplier no.:
    59005

Specifications

About this item

 :
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049