Order Entry
Puerto Rico
ContactUsLinkComponent
Calcimycin (A23187) free acid ≥98% (by TLC) ionophore
Irritant
Calcimycin (A23187) free acid ≥98% (by TLC) ionophore
Catalog # 89139-282
Supplier:  Biotium
CAS Number:  
Irritant
Calcimycin (A23187) free acid ≥98% (by TLC) ionophore
Catalog # 89139-282
Supplier:  Biotium
Supplier Number:  59001
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89139-282
  • Supplier no.:
    59001

Specifications

About this item

 :
 :523.63 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202