About this item
The Tailveiner® is the first easy-to-use mouse and rat restrainer specifically designed for tail vein injections.
- Slotted tube permits the rat or mouse to be quickly pulled in by the tail
- Suitable for intravenous injections in small animals
The animal is restrained by a plug which slides easily to accommodate any size rat or mouse, but locks automatically when handle is released. After dosing, the handle is lifted and the plug is easily removed.