ContactUsLinkComponent
Three-Way PTFE Stopcock for KIMAX® Burets, Kimble Chase
Three-Way PTFE Stopcock for KIMAX® Burets, Kimble Chase
Catalog # 89217-738
undefined
Three-Way PTFE Stopcock for KIMAX® Burets, Kimble Chase
Catalog # 89217-738
Supplier Number:  41530F-2
Specifications
  • Description:
    Three way stopcock plug
  • Cat. No.:
    89217-738
  • Bore size:
    2 mm
  • Plug size:
    12.9/44 mm
  • Supplier no.:
    41530F-2
Specifications
About this item
Three-Way PTFE stopcock plug is chemically-resistant and self-lubricating.