ESD Safe Workstation Covers, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Drape ESD workstation covers over sensitive products to protect from ESD, dust, and other contaminants.
- Groundable for added safety; 4 mm grounding snap included
- Polyester with conductive yarn
- ESD-safe logo designates safe use in ESD protective environments
- Resistance less than 10⁵
- Bound edges
ESD workstation covers create a Faraday cage effect around the covered product to shield from electrostatic fields and electrostatic discharges (ESD). This washable cover has a surface resistivity of 10⁵ ohms/sq. Covers feature a silk-screened ESD symbol.