ContactUsLinkComponent
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥98% (by TLC) fluorescent pH indicator
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥98% (by TLC) fluorescent pH indicator
  89139-254
 :  Biotium
 :  
undefined
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥98% (by TLC) fluorescent pH indicator
  89139-254
 :  Biotium
 :  51016
 :  
  • Size:
    100 mg
  • Cat. No.:
    89139-254
  • Supplier no.:
    51016
 :
 :529.28 g/mol
 :GHS07
 :Refrigerator
 :127770-45-0
 :C₂₅H₁₄Cl₂O₉
 :MFCD08459585