Order Entry
Puerto Rico
ContactUsLinkComponent
6-Methoxy-N-(3-sulfopropyl)quinolinium (SPQ) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Irritant
6-Methoxy-N-(3-sulfopropyl)quinolinium (SPQ) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 89139-272
Supplier:  Biotium
CAS Number:  
Irritant
6-Methoxy-N-(3-sulfopropyl)quinolinium (SPQ) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 89139-272
Supplier:  Biotium
Supplier Number:  52010
CAS Number:  

Specifications

  • Size:
    50 mg
  • Cat. No.:
    89139-272
  • Supplier no.:
    52010

Specifications

About this item

 :
 :281 g/mol
 :GHS07
CAS Number:83907-40-8
Formula:C₁₅H₁₅NO₄S
MDL Number:MFCD00037592