ContactUsLinkComponent
Conductive ESD Waste Basket, 7 Gallon, Transforming Technologies
Catalog # 76209-976
undefined
Conductive ESD Waste Basket, 7 Gallon, Transforming Technologies
Catalog # 76209-976
Supplier Number:  WBAS28
Specifications
  • Color:
    Black
  • Size:
    7 gal.
  • Cat. No.:
    76209-976
  • Ext. dimensions:
    26×15×11"
Specifications
About this item
Conductive ESD waste baskets remove the static threat of plastic receptacles that can damage sensitive electronic devices.
  • 7 gallon capacity
  • Permanently conductive; will never lose ESD properties
  • ESD symbol in yellow printed on body for easy identification
  • Typical point-to-point resistance: 10⁴ to 10⁵ ohm
  • Carbon loaded, black conductive polypropylene construction
ESD/anti-static trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark. When grounded, the cans and liners will not hold a static charge and they will contain materials safely. Ideal for cleanrooms, electronics manufacturing, inspection, quality control, and research laboratories. This durable and stylish rectangular wastebasket features radius corners and rolled edges. It will not dent, crush, or chip.
Certifications: Complies with IEC 61340-5-1. Meets or exceeds requirements of ANSI ESD-S20.20