Conductive ESD Trash Can, 22 Gallon, Transforming Technologies
Catalog # 76210-008
Supplier: Transforming Technologies
Specifications
- Color:Black
- Size:22 gal.
- Environmentally Preferable:
- Cat. No.:76210-008
- Ext. dimensions:30×20×11"
Specifications
About this item
Conductive ESD trash cans remove the static threat of plastic receptacles that can damage sensitive electronic devices.
- 22 gallon capacity
- Permanently conductive; will never lose ESD properties
- ESD symbol in yellow printed on body for easy identification
- Typical point-to-point resistance: 10⁴ to 10⁵ ohm
- Carbon loaded, black conductive polypropylene construction
ESD/anti-static trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark. When grounded, the cans and liners will not hold a static charge and they will contain materials safely. Ideal for electronics manufacturing, inspection, quality control, and research laboratories. This durable container is a stylish rectangular wastebasket with radius corners and rolled edges. It will not dent, crush, or chip.
Certifications: Complies with IEC 61340-5-1. Meets or exceeds requirements of ANSI ESD-S20.20