Orders LinkContactUsLinkComponent
ESD SMD Boxes, Transforming Technologies
ESD SMD Boxes, Transforming Technologies
Certificates
ESD SMD Boxes, Transforming Technologies
ESD SMD Boxes, Transforming Technologies
8 Options Available Below
Product Details & Documents
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
  • Ideal for engineering labs
  • Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
 : Meets or exceeds requirements of ANSI ESD-S20.20
Product Family Options
ESD SMD Boxes with Black Lids
Product Information
  • Ext. dimensions
    Availability
    Price
  • Small
    16×12×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $329.00
    $329.00
    Pack of 100
       $329.00
    Supplier Number: SM0873
  • Medium
    37×12×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $211.00
    $211.00
    Pack of 50
       $211.00
    Supplier Number: SM0874
  • Large
    40×37×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $126.00
    $126.00
    Pack of 25
       $126.00
    Supplier Number: SM0875
  • 76209-952
    Extra-large
    Extra-large
    68×57×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $68.50
    $68.50
    Pack of 10
       $68.50
    Supplier Number: SM0876
ESD SMD Boxes with Transparent Lids
Product Information
  • Ext. dimensions
    Availability
    Price
  • Small
    16×12×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $474.00
    $474.00
    Pack of 100
       $474.00
    Supplier Number: SM0880
  • Medium
    37×12×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $361.00
    $361.00
    Pack of 50
       $361.00
    Supplier Number: SM0881
  • Large
    40×37×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $204.00
    $204.00
    Pack of 25
       $204.00
    Supplier Number: SM0882
  • 76209-960
    Extra-large
    Extra-large
    68×57×15 mm (int)
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $96.00
    $96.00
    Pack of 10
       $96.00
    Supplier Number: SM0883