MT4000 Series Textured Two-Layer ESD Table Mats, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Mats create an ESD safe work area by a providing a path-to-ground for electrical charges, as well as protecting the work surface from damage.
- Non-slip textured surface
- Excellent resistance to hold solders, soldering irons, and most solvents
- Durable 0.080" thick two-layer rubber
The MT4000 Textured Two-Layer ESD Rubber Mats are made of 100% co-extruded rubber. They are constructed with a textured, dissipative top layer and a conductive bottom layer. The rolls are available in multiple widths.
Certifications: Meets or exceeds requirements of ANSI ESD S541-2003. RoHS compliant
Frequently Bought Together