Specifications
- Description:Ptec™ AirFLEX flexible neck ionizing nozzle with photoelectric eye
- Cat. No.:76209-932
- Electrical:100/240 V, 50/60 Hz
Specifications
About this item
The IN4200PE flexible nozzle neck frees operator’s hands during assembly and manufacturing processes. A photoelectric eye controls both ionization and airflow, which reduces compressed air costs and extends the life of the ionizer.
- Flexible neck allows users to optimally position airflow
- Hands-free operation, with a photoelectric eye
- Multiple nozzle options to customize airflow for specific applications
- Mountable
- No calibration required
The AirFLEX IN4200PE Ionizer with proximity sensor controls the power and air flow for easy hands-free precision ionization. The flexible neck allows users to target the air flow directly at an intended surface for optimal static eliminating performance. Airflow is triggered when the sensor detects a nearby object, enabling hands-free ionization only when needed. These ionizers are valuable because they can be mounted in hard to reach areas that larger ionizers just cannot handle. Emitter life is increased and maintenance frequency is reduced, due to 68 KHz AC ion emission. It is designed to neutralize electrostatic charges in benchtop environments, sensitive materials assembly, medical device, SMT, electronics assembly, automotive packaging, cleanroom, and laboratory environments.