ContactUsLinkComponent
CrossFLOW™ Wide Coverage Benchtop Ionizing Blower with Heater, Transforming Technologies
CrossFLOW™ Wide Coverage Benchtop Ionizing Blower with Heater, Transforming Technologies
Catalog # 76209-928
undefined
CrossFLOW™ Wide Coverage Benchtop Ionizing Blower with Heater, Transforming Technologies
Catalog # 76209-928
Supplier Number:  IN3000
Specifications
  • Description:
    CrossFLOW™ ionizing blower with heater
  • Cat. No.:
    76209-928
  • Electrical:
    100/240 V, 50/60 Hz
Specifications
About this item
The CrossFLOW™ ionizing blower produces a powerful air flow, rich in positive and negative ions that quickly neutralize static charges.
  • Rapid static elimination performance
  • Wide coverage range: 3×6' foot area
  • Integrated heater
  • Integrated emitter point cleaner
Powered by inherently balanced AC technology, the ionizer provides a stable output with exceptionally fast decay performance. The CrossFLOW™ features built-in emitter point cleaners and a heater that can be turned on at any time for user comfort. It has an effective range of 3×6 ft. which can protect most work areas, and features an adjustable multi-speed blower to produce a uniform air flow that can tackle any static application. Great for wide coverage applications, it can be used in benchtop environments, sensitive materials assembly, medical device, SMT, electronics assembly, automotive packaging, cleanroom, and laboratory environments.