Full Coverage ESD Sole Grounders, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Full coverage sole grounders provide 20 times the surface contact of heel grounders, with a wider and more consistent contact area to ensure grounding.
- Rapid and complete static dissipation due to constant contact through entire walking motion
- Longer life than heel grounders
Wearing the conductive ribbon inside the shoe or sock assures proper electrical contact with the user. A rugged 1 meg ohm buried resistor is standard. Heel grounders are worn on both feet to provide consistent grounding while in motion.
Certifications: Meets or exceeds requirements of ANSI ESDS20.20