Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-NM-PP1
1-NM-PP1 ≥98% (by HPLC)
1-NM-PP1 ≥98% (by HPLC)
1-NM-PP1 ≥98% (by HPLC)
Supplier:
Avantor
Certificates
Expand image
1-NM-PP1 ≥98% (by HPLC)
Supplier:
Avantor
About this item
CAS Number:
221244-14-0
:
Formula:
C₂₀H₂₁N₅
MDL Number:
MFCD17010191
:
331.41 g/mol
:
GHS06
Storage Temperature:
Freezer
Related Information
Documentation
Product specification
Spec_1-NM-PP1.pdf
Documentation