ChemMax® 3 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists, Attached Boots, Zipper Closure with Storm Flap, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Chemical Protective Coveralls are made with advanced ChemMax® 3 chemcial protective fabric for proven protection against a broad spectrum of toxic industrial chemicals, chemical warfare agents, ammonia, and fentanyl.
- ChemMax® 3 chemical protective film
- Heat sealed seams
- Respirator-fit hood
- Elastic wrists and attached boots
- Exceptional protection from Ammonia and Fentanyl
- Supported by Lakeland's PermaSURE® Chemical Toxicity Risk Modeling Software
Heat sealed seams and a storm flap over the zipper ensure maximum protection.
Specifications
- Catalog No:
- 76314-140
- 76314-142
- 76314-144
- 76314-146
- 76314-148
- 76314-150
- 76314-152
- Seam type:
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection