ChemMax® 2 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists, Attached Boots, Zipper Closure with Storm Flap, Lakeland Industries
: Lakeland
Chemical protective coveralls are made with the proven protection of Saranex® 23P barrier film for protection in Hazmat, chemical handling, and environmental cleanup applications.
- Heat sealed seams
- Respirator-fit hood
- Attached boots
ChemMax® 2 chemical protectve coveralls provide protection from chemical mixing and splash, environmental cleanup, and hazardous materials remediation. These garments feature a respirator-fit hood, attached boots, and heat sealed seams to ensure maximum protection.
- Catalog No:
- 76314-098
- 76314-096
- 76314-094
- 76314-092
- 76314-104
- 76314-102
- 76314-100
- Seam type:
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Heat sealed
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection