Order Entry
United States
ContactUsLinkComponent
Crystal violet solution reagent C for PREVI® COLOR GRAM instrument
Crystal violet solution reagent C for PREVI® COLOR GRAM instrument
Catalog # 89127-616
Supplier:  bioMerieux
CAS Number:  
Crystal violet solution reagent C for PREVI® COLOR GRAM instrument
Catalog # 89127-616
Supplier:  bioMerieux
Supplier Number:  29587
CAS Number:  

Specifications

  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    89127-616
  • Supplier no.:
    29587

Specifications

About this item

Synonyms: CI 42555
 :
 :407.99 g/mol
 :GHS02
 :GHS07
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750