Order Entry
United States
ContactUsLinkComponent
Endotoxin Removal System, EndoTrap® HD, Biovendor
Endotoxin Removal System, EndoTrap® HD, Biovendor
Catalog # 103737-146
Supplier:  BioVendor
CAS Number:  
Endotoxin Removal System, EndoTrap® HD, Biovendor
Catalog # 103737-146
Supplier:  BioVendor
Supplier Number:  LET0010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    EndoTrap® HD 5/1
  • Environmentally Preferable:
  • Cat. No.:
    103737-146

Specifications

About this item

EndoTrap® HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples.

  • Excellent recovery rate
  • Designed to meet the needs of pharma and biotech industry
  • Efficient removal of endotoxins in biomanufacturing processes

EndoTrap® HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate.

The EndoTrap HD has an endotoxin binding capacity of >5000000 EU/ml. EndoTrap removes more than 99.99% of LPS and has a sample recovery rate of >95%. The non-toxic bacteriophage-derived ligand so that there are no cytotoxic effects.

EndoTrap® HD is based on hydrophilic, dimensionally stable affinity matrix with excellent pressure/flow characteristics. A EndoTrap® Leakage ELISA is available for the quantitative determination of the EndoTrap® ligand leakage.

Ordering information: EndoTrap® HD 5/1 Endotoxin Removal System includes 5x1mL columns, 125mL 5x Equilibration Buffer, 125mL 5x Regeneration Buffer, and 125mL 5x Storage Buffer. Developed by Hyglos and manufactured by LIONEX.