ContactUsLinkComponent
Tissue Culture Plate, Midsci
Tissue Culture Plate, Midsci
Catalog # MSPP-TP92096
Supplier:  MIDSCI MS
Tissue Culture Plate, Midsci
Catalog # MSPP-TP92096
Supplier:  MIDSCI MS
Supplier Number:  TP92096
Specifications
  • Well volume:
    340 µl
  • Description:
    96 Well TC Plate
  • Cat. No.:
    MSPP-TP92096
  • Well shape:
    Flat
Specifications
About this item
The tissue culture plate has excellent well to well consistency and reduced edge effect. The black alphanumerics on top for quick and easy identification of wells and clear alphanumerics between wells on the bottom for easy orientation during microscopy, the yellow marking areas on lids and plates make labeling easy and also ensure correct lid orientation.
  • Made from optically clear polystyrene for excellent viewing
  • Sterile, packaged individually or in convenient multipacks
Each plate has a vented base that guarantees secure stacking while allowing air flow to prevent condensation and sticking of stacked plates. Textured easy-grip area ensures a secure grasp and prevents from accidentally lifting off the lid.