Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Beaker Brush, Justman

Beaker Brush, Nylon, Black, 12.7 cm (5"), 76.2 mm (3”), Wood, 41 cm (16")
  30620-600
 :  J720400
Beaker Brush, Justman
  30620-600
Beaker Brush, Justman

Beaker Brush, Nylon, Black, 12.7 cm (5"), 76.2 mm (3”), Wood, 41 cm (16")

 
$70.58
   
Each
 

Designed for heavy-duty cleaning of beakers, jars, and other wide-mouth glassware.

  • Nylon bristles are spirally wound into grooves in the smooth wood handle
  • Heavy tufting at the tip reduces scratching and improves cleaning capability
 
  • Bristle color:
    Black
  • Bristle material:
    Nylon
  • Brush head Ø:
    76.2 mm (3”)
  • Handle material:
    Wood
  • Description:
    Beaker Brush
  • Total length:
    41 cm (16")
  • Brush head length:
    12.7 cm (5")