ContactUsLinkComponent
First Strand Buffer, 5X, Teknova
Catalog # 100217-306
Supplier:  Avantor
CAS Number:  
First Strand Buffer, 5X, Teknova
Catalog # 100217-306
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    First Strand Buffer, 5X
  • Size:
    500 ml
  • Cat. No.:
    100217-306
  • Supplier no.:
    F1204
Specifications
About this item
First strand synthesis buffer designed for use in reverse transcription.
Consists of Tris-HCl (250 mM), pH 8.3, potassium chloride (375 mM), magnesium chloride (15 mM) and DTT (50 mM).