Order Entry
Puerto Rico
ContactUsLinkComponent
Normal Swine Serum, Jackson Immunoresearch
Normal Swine Serum, Jackson Immunoresearch
Catalog # 102643-880
Normal Swine Serum, Jackson Immunoresearch
Catalog # 102643-880
Supplier Number:  014-000-121

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Serum format:
    Lyophilized
  • Description:
    Normal Swine Serum
  • Size:
    10 ml
  • Storage Temperature:
    2 - 8 °C
  • Concentration:
    60 mg/ml
  • Cat. No.:
    102643-880
  • Supplier no.:
    014-000-121

Specifications

About this item

Normal serums are lipid extracted to improve clarity.

  • Dialyzed against PBS containing sodium azide
  • Freeze-dried
  • Strongly recommended blocking agent

Normal serum diluted to 5% (v/v) in PBS to reduce background from non-specific, conserved sequence and/or Fc receptor binding. Best results are obtained with diluted normal serum from the same host as the labeled antibody, as a separate incubation step before addition of the primary antibody.