Anti-HA.11 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16B12]
Supplier: Biolegend
Certificates
About this item
The HA tag (hemagglutinin) is an amino acid sequence derived from the human influenza hemagglutinin surface glycoprotein, corresponding to amino acids 98 to 106.
- Formulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide
- Excitation laser: Blue laser (488 nm), green laser (532 nm)/yellow-green laser (561 nm)
It is commonly used as a tag to facilitate detection, isolation, and purification of proteins.
Preparation: The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Type: Tag
Antigen: HA.11
Clonality: Monoclonal
Clone: 16B12
Conjugation: PE (Phycoerythrin)
Epitope: HA
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 76302-662
- 76302-664
- Antigen Symbol:
- HA.11
- HA.11
- Antigen Name:
- Hemagglutin 11
- Hemagglutin 11
- Antibody Type:
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 16B12
- 16B12
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Monoclonal Antibody HA.11 was Raised Against the Twelve Amino Acid Peptide CYPYDVPDYASL
- Monoclonal Antibody HA.11 was Raised Against the Twelve Amino Acid Peptide CYPYDVPDYASL
- Sequence:
- YPYDVPDYA
- YPYDVPDYA
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C