NMR Sample Tube Screw Caps and Septa, New Era™
Supplier: New ERA Enterprises
Certificates
About this item
Open-top caps with PTFE/silicone septum combine to provide the most inert closure with needle access.
Ordering information: PTFE/silicone septa are also available separately.