Thin Walled Series NMR Sample Tubes, 10 mm, New Era™
All large volume NMR sample tubes are made of type 1, class A borosilicate glass (Pyrex or equivalent), unless otherwise noted.
- Round bottom is standard
- Flat bottoms are available upon request
All large volume NMR sample tubes are made of type 1, class A borosilicate glass (Pyrex or equivalent), unless otherwise noted.