PVC Microbore Tubing Links, Cole Parmer
Supplier: Avantor
Certificates
About this item
Tubing made of PVC based material with plasticizer.
- Thermoplastic
- Firm stiff material
- Transparent
Temperature resistance from −50 to 74 °C.
Certifications: Conforms to FDA 21 CFR 175.300.