Order Entry
Puerto Rico
ContactUsLinkComponent
Biocytin TMR, CF® 640R Dye
Biocytin TMR, CF® 640R Dye
Catalog # 75784-060
Supplier:  Biotium
CAS Number:  
Biocytin TMR, CF® 640R Dye
Catalog # 75784-060
Supplier:  Biotium
Supplier Number:  92007
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    0.5 mg
  • Cat. No.:
    75784-060
  • Supplier no.:
    92007

Specifications

Specification Test Results

  • Abs/Em Maxima:
    642/662 nm
  • Extinction coefficient:
    105,000
  • Molecular weight:
    ~ 832
  • Excitation laser line:
    633 nm, 635 nm or 640 nm
  • Replaces:
    Alexa Fluor® 647, ATTO 647N, Cy®5, DyLight® 649

Specification Test Results

About this item

 :
 :869.08 g/mol
Formula:C₄₆H₈₂N₈O₇S