Order Entry
Puerto Rico
ContactUsLinkComponent
Biocytin TMR, CF® 594 Dye
Biocytin TMR, CF® 594 Dye
Catalog # 75784-058
Supplier:  Biotium
CAS Number:  
Biocytin TMR, CF® 594 Dye
Catalog # 75784-058
Supplier:  Biotium
Supplier Number:  92006
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    0.5 mg
  • Cat. No.:
    75784-058
  • Supplier no.:
    92006

Specifications

Specification Test Results

  • Abs/Em Maxima:
    593/614 nm
  • Extinction coefficient:
    115,000
  • Molecular weight:
    ~730
  • Excitation laser line:
    532 nm, 568 nm or 594 nm
  • Replaces:
    Alexa Fluor® 594, DyLight® 594, Texas Red®

Specification Test Results

About this item

 :
 :869.08 g/mol
Formula:C₄₆H₈₂N₈O₇S