Biocytin TMR, CF® 488A Dye
Catalog # 75784-038
Supplier: Biotium
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:0.5 mg
- Cat. No.:75784-038
- Supplier no.:92004
Specifications
Specification Test Results
- Abs/Em Maxima:490/515 nm
- Extinction coefficient:70,000
- Molecular weight:~914
- Excitation laser line:488 nm
- Replaces:Alexa Fluor® 488, DyLight® 488, fluorescein (aka FITC, FAM), Cy®2
Specification Test Results
About this item
:
:869.08 g/mol
Formula:C₄₆H₈₂N₈O₇S