EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98% (by complexometric analysis, on dry substance), Molecular Biology Grade, Calbiochem®
Catalog # 80502-396
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:25 g
- Cat. No.:80502-396
- Supplier no.:324626-25GM
Specifications
Specification Test Results
- Purity (Complexometry, Dry Basis):≥98%
- Heavy Metals:≤0.001% (as Pb)
- Contaminants (DNases, proteases, RNases):none detected
Specification Test Results
About this item
:
Merck Index:13,03560
Melting Pt:240…244 °C
:380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291