Versaflo™ Single Station Battery Charger Kit, 3M™
76312-268
: 3M
- Description:Versaflo™ single station battery charger kit
- Cat. No.:76312-268
This battery charger kit is a complete single station battery charger for TR-600 series PAPRs. It consists of a cradle and a power supply.
- Easy to use
- Quickly charging batteries
- Holds charged batteries ready for use
- Charging time: less than four hours
This battery charger will charge a standard capacity battery or high capacity battery in less than four hours. It will charge one battery at a time. Batteries may be stored on the charger, fully charged, ready for immediate use.
: NIOSH approved.