Bacterial Test Standard (BTS), Bruker
Catalog # MSPP-8255343
Supplier: BRUKER SCIENTIFIC, LLC MS
CAS Number:
Specifications
- Description:Bacterial Test Standard
- Cat. No.:MSPP-8255343
- Supplier no.:8255343
Specifications
About this item
This bacterial test standard (BTS) is used in the MALDI Biotyper system quality control process to ensure reliable and accurate identification of microorganisms.
Consisting of an E. coli extract spiked with two high-molecular–weight proteins, its composition covers the entire mass range of proteins
measured during MALDI Biotyper identifications.
Delivery: Box containing five tubes, each sufficient for approximately 40 sample positions.