Metal Needle Kit for 32-Gauge Stainless Steel Needle, Thermo Electron
MSPP-OPTON5300
- Description:Metal needle kit, OPTON-53003
- Cat. No.:MSPP-OPTON5300
- Supplier no.:OPTON-53003
The kit contains ferrule (0.012" I-Ø), clear; external union, 10-32, natural PEEK; Internal union, 10-32, natural PEEK; 10-32×¼-28, Kel-F, adapter for sample inlet of ESI probe; high-pressure, fingertight nut and ferrule for ₁/₁₆" I-Ø tubing; stainless steel needle, 32-gauge, blunt-tipped, 5.65" length, 0.004×0.009" I-Ø; 0.005" I-Ø, red PEEK tubing.
: The kit includes 6 ferrule, 2 external union, 1 internal union, 1 Kel-F adapter, 2 high-pressure fingertight nut and ferrule and 3 stainless steel needle.