Vi-CELL™ MetaFLEX Sensor Cassette, Beckman Coulter
Catalog # MSPP-B93030
Supplier: Beckman Coulter
Specifications
- Description:Vi-CELL™ MetaFLEX Sensor Cassette 300/30, White
- Cat. No.:MSPP-B93030
Specifications
About this item
The Vi-CELL MetaFLEX sensor cassette SC300 measures pH, pO2, pCO2, Glucose, Lactate, Electrolytes and more parameters associated with whole blood analysis.
For use with the Vi-CELL MetaFLEX™ bioanalyte analyser.