Anti-IgG Sheep Polyclonal Antibody
Supplier: Innovative Research Inc
Certificates
About this item
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- 103387-834
- 103386-156
- Antigen Symbol:
- IgG
- IgG
- Antibody Type:
- Control
- Control
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Sheep
- Sheep
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Concentration:
- 5.0 mg/ml
- 5.0 mg/ml
- Storage Buffer:
- 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4
- 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C