Anti-REN Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat;Mouse
- Catalog No:
- 103387-816
- 103386-138
- Antigen Symbol:
- REN
- REN
- Antigen Name:
- Prorenin
- Prorenin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Rat, Mouse
- Rat, Mouse
- Host:
- Sheep
- Sheep
- Gene ID:
- 24715
- 24715
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant full length rat protein fused to a His tag at C-terminal end (Glycosylated).
- Recombinant full length rat protein fused to a His tag at C-terminal end (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 44 kDa
- 44 kDa
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C; Protect from light
- –70 °C; Protect from light
- Shipping Temperature:
- –70 °C
- –70 °C