Anti-PLAU Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Type: Primary
Antigen: PLAU
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
- Catalog No:
- 103386-130
- 103387-808
- Antigen Symbol:
- PLAU
- PLAU
- Antigen Name:
- Urokinase
- Urokinase
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Sheep
- Sheep
- Gene ID:
- 100008633
- 100008633
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant full length wild-type rabbit protein (Glycosylated).
- Recombinant full length wild-type rabbit protein (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 44 kDa
- 44 kDa
- Concentration:
- 2.0 mg/ml
- 2.0 mg/ml
- Storage Buffer:
- PBS, pH 7.4; 1% BSA, 0.01% Thimerosal
- PBS, pH 7.4; 1% BSA, 0.01% Thimerosal
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C