Anti-FGA Sheep Polyclonal Antibody
Supplier: Molecular Innovations
Certificates
About this item
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
Specifications
- Catalog No:
- 103386-110
- 103387-788
- Antigen Symbol:
- FGA
- FGA
- Antigen Name:
- Fibrinogen
- Fibrinogen
- Antigen Synonyms:
- FGB, FGG
- FGB, FGG
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Sheep
- Sheep
- Gene ID:
- 100353616, 100351341, 10035159
- 100353616, 100351341, 10035159
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Full length native protein (purified) (Rabbit).
- Full length native protein (purified) (Rabbit).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 340 kDa
- 340 kDa
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.01M Hepes; 0.15M NaCl; 50% Glycerol; pH 7.4
- 0.01M Hepes; 0.15M NaCl; 50% Glycerol; pH 7.4
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C