Anti-F10 Sheep Polyclonal Antibody
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103388-794
- 103386-022
- Antigen Symbol:
- F10
- F10
- Antigen Name:
- Factor X
- Factor X
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Sheep
- Sheep
- Gene ID:
- 14058
- 14058
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Full length native protein (purified) (Mouse).
- Full length native protein (purified) (Mouse).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 58.9 kDa
- 58.9 kDa
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C