Anti-SERPINA10 Sheep Polyclonal Antibody
Supplier: Molecular Innovations
Certificates
About this item
Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 103385-992
- 103387-694
- Antigen Symbol:
- SERPINA10
- SERPINA10
- Antigen Name:
- Protein Z-Dependent Protease Inhibitor
- Protein Z-Dependent Protease Inhibitor
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Sheep
- Sheep
- Gene ID:
- 51156
- 51156
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant full length wild-type human protein (Glycosylated).
- Recombinant full length wild-type human protein (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 72 kDa (Plasma), 50 kDa (Recombinant)
- 72 kDa (Plasma), 50 kDa (Recombinant)
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C