Anti-REN Mouse Monoclonal Antibody [clone: 4B5-E3] (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: 4B5-E3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: 30—43
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
- Catalog No:
- 103385-654
- 103387-396
- Antigen Symbol:
- REN
- REN
- Antigen Name:
- Prorenin
- Prorenin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 4B5-E3
- 4B5-E3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 5972
- 5972
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- 30—43
- 30—43
- Immunogen:
- Recombinant full length wild-type human protein (Glycosylated).
- Recombinant full length wild-type human protein (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 43.725 kDa
- 43.725 kDa
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C; Protect from light
- –70 °C; Protect from light
- Shipping Temperature:
- –70 °C
- –70 °C