Anti-KNG1 Mouse Monoclonal Antibody [clone: 17E10-G9] (Biotin)
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 17E10-G9
Conjugation: Biotin
Epitope: Light Chain
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Specifications
- Catalog No:
- 103387-074
- 103385-404
- Antigen Symbol:
- KNG1
- KNG1
- Antigen Name:
- Kininogen
- Kininogen
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- 17E10-G9
- 17E10-G9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3827
- 3827
- Isotype:
- IgG2b
- IgG2b
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Light Chain
- Light Chain
- Immunogen:
- Full length native protein (purified) (Human).
- Full length native protein (purified) (Human).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 120 kDa
- 120 kDa
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C