About this item
Type: Control
Antigen: IgG Fc
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG Fc
Reactivity:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 103385-298
- 103386-992
- Antigen Symbol:
- IgG Fc
- IgG Fc
- Antibody Type:
- Control
- Control
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Human
- Human
- Isotype:
- IgG Fc
- IgG Fc
- Form:
- Frozen liquid
- Frozen liquid
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.2M NaCl; 0.05% Azide; pH 8.0
- 0.05M Sodium Phosphate; 0.2M NaCl; 0.05% Azide; pH 8.0
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C