Anti-F11 Mouse Monoclonal Antibody [clone: 2B11-E5F6]
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: F11
Clonality: Monoclonal
Clone: 2B11-E5F6
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 103386-644
- 103384-918
- Antigen Symbol:
- F11
- F11
- Antigen Name:
- Factor XI
- Factor XI
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2B11-E5F6
- 2B11-E5F6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2160
- 2160
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Heavy Chain
- Heavy Chain
- Immunogen:
- Full length native protein (purified) (Human).
- Full length native protein (purified) (Human).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 160 kDa
- 160 kDa
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C